Novus Biologicals
Manufacturer Code:NBP274444IR
Catalog # NBP274444IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H6 |
Immunogen | Full length recombinant protein of human TASP1 (NP_060184) produced in HEK293T cells. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: C20orf13taspase 1 chromosome 20 open reading frame 13 dJ585I14.2 EC 3.4.25.- FLJ20212 MGC39159 taspase threonine aspartase 1 Taspase-1 threonine aspartase 1; Taspase-1; Threonine aspartase 1; Threonine aspartase subunit alpha; Threonine aspartase subunit beta
Gene Aliases: C20orf13; dJ585I14.2; TASP1
UniProt ID: (Human) Q9H6P5
Entrez Gene ID: (Human) 55617
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.