Invitrogen
Manufacturer Code:PA556567
Catalog # PIPA556567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TASP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Taspase threonine aspartase 1 C20orf13 FLJ20212 MGC39159 dJ585I14.2; Taspase-1; Threonine aspartase 1; Threonine aspartase subunit alpha; Threonine aspartase subunit beta
Gene Aliases: C20orf13; dJ585I14.2; TASP1
UniProt ID: (Human) Q9H6P5
Entrez Gene ID: (Human) 55617
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.