Invitrogen
Manufacturer Code:PA567753
Catalog # PIPA567753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human TAS2R9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R9; Taste receptor family B member 6; Taste receptor type 2 member 9; Taste receptor type 2 member 9 T2R9 Taste receptor family B member 6 TRB6; taste receptor, family B, member 6; taste receptor, type 2, member 9; TRB6
Gene Aliases: T2R9; TAS2R9; TRB6
UniProt ID: (Human) Q9NYW1
Entrez Gene ID: (Human) 50835
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.