Invitrogen
Manufacturer Code:PA599930
Catalog # PIPA599930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAS2R60 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC119154 MGC119155 T2R56 T2R60 TAS2R60 taste 2 receptor member 60 taste receptor type 2 member 56 Taste receptor type 2 member 60 taste receptor type 2 member 60; T2R56; T2R60; Taste receptor type 2 member 56; Taste receptor type 2 member 60; taste receptor, type 2, member 60
Gene Aliases: T2R56; T2R60; TAS2R60
UniProt ID: (Human) P59551
Entrez Gene ID: (Human) 338398
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.