Invitrogen
Manufacturer Code:PA5102256
Catalog # PIPA5102256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAS2R5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R5; T2R5 TAS2R5 taste 2 receptor member 5 taste receptor type 2 member 5 taste receptor type 2 member 5; Taste receptor type 2 member 5; taste receptor, type 2, member 5
Gene Aliases: T2R5; TAS2R5
UniProt ID: (Human) Q9NYW4
Entrez Gene ID: (Human) 54429
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.