Invitrogen
Manufacturer Code:OSR00174W
Catalog # PIOSR00174W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from human taste receptor type 2 member 47 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R46; T2R54; Taste receptor type 2 member 46; Taste receptor type 2 member 47 TAS2R47; Taste receptor type 2 member 54; taste receptor, type 2, member 46
Gene Aliases: T2R46; T2R54; TAS2R46
UniProt ID: (Human) P59540
Entrez Gene ID: (Human) 259292
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.