Novus Biologicals
Manufacturer Code:NB11074910
Catalog # NB11074910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide as a part of human taste receptor type 2 member 46 (TAS2R46 T2R46 T2R54) conjugated to immunogenic carrier protein was used as the immunogen. The antibodies generated using this antigen is expected to also recognize gene products of TAS2R43 TAS2R44 TAS2R47 and TAS2R49. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC134865 T2R54T2R46Taste receptor type 2 member 54 taste receptor type 2 member 46 taste receptor type 2 member 46; T2R46; T2R54; Taste receptor type 2 member 46; Taste receptor type 2 member 54; taste receptor, type 2, member 46
Gene Aliases: T2R46; T2R54; TAS2R46
UniProt ID: (Human) P59540
Entrez Gene ID: (Human) 259292
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.