Invitrogen
Manufacturer Code:PA5103257
Catalog # PIPA5103257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAS2R43 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R43; T2R43 T2R52 TAS2R43 taste 2 receptor member 43 taste receptor type 2 member 43 Taste receptor type 2 member 52 taste receptor type 2 member 43; T2R52; Taste receptor type 2 member 43; Taste receptor type 2 member 52; taste receptor, type 2, member 43
Gene Aliases: T2R43; T2R52; TAS2R43
UniProt ID: (Human) P59537
Entrez Gene ID: (Human) 259289
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.