Novus Biologicals
Manufacturer Code:NBP183154
Catalog # NBP183154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SLVRHTRNLKLSSLGSRDSSTEAHRRAMK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: candidate taste receptor hT2R55; hT2R55 T2R24 T2R55T2R42 TAS2R55candidate taste receptor hT2R55 taste receptor T2R55 taste receptor type 2 member 42 Taste receptor type 2 member 55 taste receptor type 2 member 42; taste receptor type 2 member 55; taste receptor, type 2, member 42
Gene Aliases: T2R24; T2R42; T2R55; TAS2R55
Entrez Gene ID: (Human) 353164
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.