Invitrogen
Manufacturer Code:PA5104309
Catalog # PIPA5104309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAS2R41 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC119928 MGC119929 mGR26 mt2r35 T2R12 T2R26 T2R41 T2R59 Tas2r12 Tas2r126 Tas2r26 Tas2r41 taste 2 receptor member 41 taste receptor rT2R12 taste receptor type 2 member 41 Taste receptor type 2 member 59 taste receptor type 2 member 126 taste receptor type 2 member 41; T2R12; T2R26; T2R41; T2R59; taste receptor rT2R12; Taste receptor type 2 member 41; Taste receptor type 2 member 59; taste receptor, type 2, member 41
Gene Aliases: mGR26; mt2r35; T2R12; T2R26; T2R41; T2R59; Tas2r12; Tas2r126; Tas2r26; TAS2R41
UniProt ID: (Human) P59536, (Rat) Q9JKE7, (Mouse) P59532
Entrez Gene ID: (Human) 259287, (Rat) 246219, (Mouse) 387353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.