Invitrogen
Manufacturer Code:PA567749
Catalog # PIPA567749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human TAS2R40. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 60; G-protein coupled receptor 60; T2R18; T2R40; T2R58; tas2r40 {ECO:0000250|UniProtKB:P59535}; Taste receptor type 2 member 18; Taste receptor type 2 member 40; Taste receptor type 2 member 40 T2R40 T2R58 G-protein coupled receptor 60 MGC126844 MGC126846; Taste receptor type 2 member 58; taste receptor, type 2, member 40
Gene Aliases: GPR60; mt2r33; RGD1562314; T2r18; T2r33; T2R40; T2R58; Tas2r144; TAS2R40; Tas2r44
UniProt ID: (Human) P59535, (Rat) Q67ET2, (Mouse) Q7TQB8
Entrez Gene ID: (Human) 259286, (Rat) 500101, (Mouse) 387515
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.