Invitrogen
Manufacturer Code:OSR00164W
Catalog # PIOSR00164W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from human taste receptor type 2 member 40 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 60; G-protein coupled receptor 60; T2R40; T2R58; Taste receptor type 2 member 40; Taste receptor type 2 member 40 T2R40 T2R58 G-protein coupled receptor 60 MGC126844 MGC126846; Taste receptor type 2 member 58; taste receptor, type 2, member 40
Gene Aliases: GPR60; T2R40; T2R58; TAS2R40
UniProt ID: (Human) P59535
Entrez Gene ID: (Human) 259286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.