Novus Biologicals
Manufacturer Code:NBP232560
Catalog # NBP232560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: ICTVYCNNSFPIHSSNSTKKTYLSEINVVGL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R39; T2R39 T2R57 taste receptor type 2 member 39 Taste receptor type 2 member 57 taste receptor type 2 member 39; T2R57; Taste receptor type 2 member 39; Taste receptor type 2 member 57; taste receptor, type 2, member 39
Gene Aliases: T2R39; T2R57; TAS2R39
UniProt ID: (Human) P59534
Entrez Gene ID: (Human) 259285
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.