Invitrogen
Manufacturer Code:PA599928
Catalog # PIPA599928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAS2R38 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mT2R31; mT2R31 PTC PTC bitter taste receptor RGD:1560649} T2R138 T2R26 T2r31 T2r38 T2r38 {ECO:0000312 T2R61 Tas2r138 Tas2r26 TAS2R38 taste 2 receptor member 38 taste receptor type 2 member 26 Taste receptor type 2 member 38 taste receptor type 2 member 61 taste receptor type 2 member 138 taste receptor type 2 member 38; PTC bitter taste receptor; T2R138; T2R26; T2R38; T2r38 {ECO:0000312|RGD:1560649}; T2R61; Taste receptor type 2 member 26; Taste receptor type 2 member 38; Taste receptor type 2 member 61; taste receptor, type 2, member 38
Gene Aliases: mt2r31; PTC; T2R138; T2r31; T2R38; T2R61; Tas2r138; Tas2r26; TAS2R38
UniProt ID: (Human) P59533, (Mouse) Q7TQA6, (Rat) Q4VHE7
Entrez Gene ID: (Human) 5726, (Mouse) 387513, (Rat) 500091
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.