Invitrogen
Manufacturer Code:PA562953
Catalog # PIPA562953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TAS2R38 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PTC bitter taste receptor; T2R38; T2R61; Taste receptor type 2 member 38; Taste receptor type 2 member 38 T2R38 T2R61 PTC bitter taste receptor PTC; Taste receptor type 2 member 61; taste receptor, type 2, member 38
Gene Aliases: PTC; T2R38; T2R61; TAS2R38
UniProt ID: (Human) P59533
Entrez Gene ID: (Human) 5726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.