Novus Biologicals
Manufacturer Code:NBP29516820UL
Catalog # NB024499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 150-250 of human TAS2R38 (NP_789787.4). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PTC bitter taste receptor; PTCphenylthiocarbamide tasting T2R38 T2R61PTC bitter taste receptor taste receptor type 2 member 38 Taste receptor type 2 member 61 taste receptor type 2 member 38; T2R38; T2R61; Taste receptor type 2 member 38; Taste receptor type 2 member 61; taste receptor, type 2, member 38
Gene Aliases: PTC; T2R38; T2R61; TAS2R38
UniProt ID: (Human) P59533
Entrez Gene ID: (Human) 5726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.