Invitrogen
Manufacturer Code:PA567773
Catalog # PIPA567773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human TAS2R30. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R30; T2R47; taste receptor T2R47; Taste receptor type 2 member 30; Taste receptor type 2 member 44 T2R44 T2R53 TAS2R44; Taste receptor type 2 member 47; taste receptor, type 2, member 30; taste receptor, type 2, member 47; type 2 taste receptor member 30; type 2 taste receptor member 47
Gene Aliases: T2R30; T2R47; TAS2R30; TAS2R47
UniProt ID: (Human) P59541
Entrez Gene ID: (Human) 259293
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.