Abnova
Manufacturer Code:H00050840K
Catalog # 89-202-315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TAS2R14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TAS2R14 peptide using ARM Technology.
For Research Use Only
Protein Aliases: MGC125491 MGC125492 T2R14 TRB1; T2R14; Taste receptor family B member 1; Taste receptor type 2 member 14; taste receptor, family B, member 1; taste receptor, type 2, member 14; TRB1
Gene Aliases: T2R14; TAS2R14; TRB1
UniProt ID: (Human) Q9NYV8
Entrez Gene ID: (Human) 50840
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.