Invitrogen
Manufacturer Code:OSR00160W
Catalog # PIOSR00160W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from aa region 210-260 of human taste receptor type 2 member 13. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R13; Taste receptor family B member 3; Taste receptor type 2 member 13; Taste receptor type 2 member 13 T2R13 Taste receptor family B member 3 TRB3; taste receptor, family B, member 3; taste receptor, type 2, member 13; TRB3
Gene Aliases: T2R13; TAS2R13; TRB3
UniProt ID: (Human) Q9NYV9
Entrez Gene ID: (Human) 50838
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.