Invitrogen
Manufacturer Code:PA539708
Catalog # PIPA539708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human TAS2R10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R10; Taste receptor family B member 2; Taste receptor type 2 member 10; Taste receptor type 2 member 10 T2R10 Taste receptor family B member 2 TRB2 MGC126811 MGC126813; taste receptor, family B, member 2; taste receptor, type 2, member 10; TRB2
Gene Aliases: T2R10; TAS2R10; TRB2
UniProt ID: (Human) Q9NYW0
Entrez Gene ID: (Human) 50839
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.