Novus Biologicals
Manufacturer Code:NBP294569100UL
Catalog # NB023662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAS2R10 (NP_076410.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC126811 T2R10 T2R10 MGC126813 Taste receptor family B member 2 taste receptor family B member 2 taste receptor type 2 member 10 TRB2taste receptor type 2 member 10; T2R10; Taste receptor family B member 2; Taste receptor type 2 member 10; taste receptor, family B, member 2; taste receptor, type 2, member 10; TRB2
Gene Aliases: T2R10; TAS2R10; TRB2
UniProt ID: (Human) Q9NYW0
Entrez Gene ID: (Human) 50839
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.