Invitrogen
Manufacturer Code:PA562509
Catalog # PIPA562509
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TAS2R1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T2R1; Taste receptor family B member 7; Taste receptor type 2 member 1; Taste receptor type 2 member 1 T2R1 Taste receptor family B member 7 TRB7 TAS2R1 TRB7 MGC126778 MGC126780; taste receptor, family B, member 7; taste receptor, type 2, member 1; TRB7
Gene Aliases: T2R1; TAS2R1; TRB7
UniProt ID: (Human) Q9NYW7
Entrez Gene ID: (Human) 50834
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.