Abnova
Manufacturer Code:PAB25087
Catalog # 89-126-761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to TAS2R1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TAS2R1.
For Research Use Only
Protein Aliases: MGC126778; MGC126780; T2R1; Taste receptor family B member 7; Taste receptor type 2 member 1; taste receptor, family B, member 7; taste receptor, type 2, member 1; TRB7
Gene Aliases: T2R1; TAS2R1; TRB7
UniProt ID: (Human) Q9NYW7
Entrez Gene ID: (Human) 50834
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.