Abnova
Manufacturer Code:H00050834K
Catalog # 89-204-607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TAS2R1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TAS2R1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: MGC126778 MGC126780 T2R1 TRB7; T2R1; Taste receptor family B member 7; Taste receptor type 2 member 1; taste receptor, family B, member 7; taste receptor, type 2, member 1; TRB7
Gene Aliases: T2R1; TAS2R1; TRB7
UniProt ID: (Human) Q9NYW7
Entrez Gene ID: (Human) 50834
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.