Novus Biologicals
Manufacturer Code:NBP233683
Catalog # NBP233683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: WLTSDLVMGLPGMAQMGTVLGFLQRGAQLHEFPQYVKTHLALATDPAFCSALGEREQGLEEDVVGQRCPQCDCITLQNVSAGLNHHQTFSVYAA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sweet taste receptor T1R3; T1R3Sweet taste receptor T1R3 taste receptor type 1 member 3 taste receptor type 1 member 3 TR3; Taste receptor type 1 member 3; taste receptor, type 1, member 3
Gene Aliases: T1R3; TAS1R3; TR3
UniProt ID: (Human) Q7RTX0
Entrez Gene ID: (Human) 83756
Molecular Function:
G-protein coupled receptor
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.