Invitrogen
Manufacturer Code:PA534254
Catalog # PIPA534254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from N-terminal extracellular domain of human TAS1R2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 71; G-protein coupled receptor 71; Sweet taste receptor T1R2; TAS1R2 G protein-coupled receptor 71 GPR71 GPCRB4 T1R2 Taste receptor type 1 member 2 TR2 Sweet taste receptor T1R2 G-protein coupled receptor 71; Taste receptor type 1 member 2; taste receptor, type 1, member 2
Gene Aliases: GPR71; T1R2; TAS1R2; TR2
UniProt ID: (Human) Q8TE23
Entrez Gene ID: (Human) 80834
Molecular Function:
G-protein coupled receptor
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.