Invitrogen
Manufacturer Code:PA5116736
Catalog # PIPA5116736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 450-550 of human TAS1R2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 4 family member A1 Aldh4a1 candidate taste receptor T1R2 G protein-coupled receptor 71 GPCRB4 GPR71 G-protein coupled receptor 71 sweet taste receptor T1R2 T1r2 Tas1r2 taste 1 receptor member 2 taste receptor TR2 taste receptor type 1 member 2 taste receptor type 1 member 2 TR2 TS1R2; G protein-coupled receptor 71; G-protein coupled receptor 71; Sweet taste receptor T1R2; Taste receptor type 1 member 2; taste receptor, type 1, member 2
Gene Aliases: GPR71; T1R2; TAS1R2; TR2
UniProt ID: (Human) Q8TE23
Entrez Gene ID: (Human) 80834
Molecular Function: G-protein coupled receptor ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.