Invitrogen
Manufacturer Code:OSR00182W
Catalog # PIOSR00182W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 1t extracellular domain of mouse taste receptor type 1 member 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: candidate taste receptor T1R1; G protein-coupled receptor 70; G-protein coupled receptor 70; Taste receptor type 1 member 1; Taste receptor type 1 member 1 (precursor) Tas1r1 Gpr70 T1r1 Tr1 TS1R1
Gene Aliases: Gpr70; T1r1; Tas1r1; TR1
UniProt ID: (Mouse) Q99PG6
Entrez Gene ID: (Mouse) 110326
Molecular Function:
G-protein coupled receptor
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.