Abnova
Manufacturer Code:H00080835A01
Catalog # 89-107-382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 281 a.a. &bsim 380 a.a) partial recombinant protein with GST tag.,TAS1R1 (NP_619642 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant TAS1R1.
For Research Use Only
Protein Aliases: G protein-coupled receptor 70; G-protein coupled receptor 70; gm148; GPR70; seven transmembrane helix receptor; T1R1; Taste receptor type 1 member 1; taste receptor, type 1, member 1; TR1
Gene Aliases: GM148; GPR70; T1R1; TAS1R1; TR1
UniProt ID: (Human) Q7RTX1
Entrez Gene ID: (Human) 80835
Molecular Function:
G-protein coupled receptor
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.