Invitrogen
Manufacturer Code:PA572547
Catalog # PIPA572547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 638-665 amino acids from the C-terminal region of human TARSL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: probable threonine--tRNA ligase 2 cytoplasmic probable threonyl-tRNA synthetase 2 cytoplasmic thrRS threonine--tRNA ligase threonyl-tRNA synthetase-like protein 2; probable threonyl-tRNA synthetase 2, cytoplasmic; threonine--tRNA ligase; Threonine--tRNA ligase 2, cytoplasmic; Threonyl-tRNA synthetase; Threonyl-tRNA synthetase protein 3; threonyl-tRNA synthetase-like 2; threonyl-tRNA synthetase-like protein 2; ThrRS
Gene Aliases: TARS3; TARSL2
UniProt ID: (Human) A2RTX5
Entrez Gene ID: (Human) 123283
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.