Invitrogen
Manufacturer Code:PA546633
Catalog # PIPA546633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human TARSL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: probable threonine--tRNA ligase 2, cytoplasmic; Probable threonyl-tRNA synthetase 2, cytoplasmic; threonine--tRNA ligase; Threonine--tRNA ligase 2, cytoplasmic; Threonyl-tRNA synthetase; threonyl-tRNA synthetase like 2; Threonyl-tRNA synthetase protein 3; threonyl-tRNA synthetase-like 2; threonyl-tRNA synthetase-like protein 2; ThrRS
Gene Aliases: A530046H20Rik; RGD1311308; si:zfos-508h5.3; TARS3; TARSL2
UniProt ID: (Bovine) A6QNM8, (Human) A2RTX5, (Mouse) Q8BLY2
Entrez Gene ID: (Rabbit) 100346648, (Bovine) 525350, (Horse) 100057955, (Dog) 479019, (Human) 123283, (Guinea pig) 100717860, (Rat) 308701, (Mouse) 272396, (Zebrafish) 100330276
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.