Novus Biologicals
Manufacturer Code:NBP293450100UL
Catalog # NB028776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human TARS2 (NP_079426.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.1.1.3 mitochondrial TARSL1 Threonine--tRNA ligase threonyl-tRNA synthetase 2 mitochondrial (putative) threonyl-tRNA synthetase mitochondrial ThrRS; Threonine--tRNA ligase, mitochondrial; Threonyl-tRNA synthetase; Threonyl-tRNA synthetase-like 1; ThrRS
Gene Aliases: COXPD21; TARS2; TARSL1; thrRS
UniProt ID: (Human) Q9BW92
Entrez Gene ID: (Human) 80222
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.