Invitrogen
Manufacturer Code:PA530690
Catalog # PIPA530690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 346 and 657 of Human ThrRS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: threonine tRNA ligase 1, cytoplasmic; Threonine--tRNA ligase 1, cytoplasmic; Threonine--tRNA ligase, cytoplasmic; Threonyl-tRNA synthetase; Threonyl-tRNA synthetase 1; threonyl-tRNA synthetase, cytoplasmic; ThrRS; ThrRS threonine tRNA ligase 1 cytoplasmic threonine--tRNA ligase cytoplasmic threonyl-tRNA synthetase cytoplasmic
Gene Aliases: D15Wsu59e; TARS; TARS1; ThrRS
UniProt ID: (Human) P26639, (Mouse) Q9D0R2
Entrez Gene ID: (Human) 6897, (Mouse) 110960
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.