Novus Biologicals
Manufacturer Code:NBP155369
Catalog # NBP155369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to TARS(threonyl-tRNA synthetase) The peptide sequence was selected from the N terminal of TARS. Peptide sequence PEYIYTRLEMYNILKAEHDSILAEKAEKDSKPIKVTLPDGKQVDAESWKT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytoplasmic EC 6.1.1.3 MGC9344 Threonine--tRNA ligase threonyl-tRNA synthetase threonyl-tRNA synthetase cytoplasmic; threonine tRNA ligase 1, cytoplasmic; Threonine--tRNA ligase 1, cytoplasmic; Threonyl-tRNA synthetase; Threonyl-tRNA synthetase 1; threonyl-tRNA synthetase, cytoplasmic; ThrRS
Gene Aliases: TARS; TARS1; ThrRS
UniProt ID: (Human) P26639
Entrez Gene ID: (Human) 6897
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.