Invitrogen
Manufacturer Code:700655
Catalog # PI700655
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | B22H33L5 |
Immunogen | A portion of recombit human TARC protein corresponding to amino acids 2494 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 17; CC chemokine TARC; CCL17 Chemokine C-C motif ligand 17 small inducible cytokine subfamily A Cys-Cys member 17 SCYA17 thymus and activation-regulated chemokine TARC ABCD-2 A-152E5.3; chemokine (C-C motif) ligand 17; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; Small-inducible cytokine A17; T cell-directed CC chemokine; Thymus and activation-regulated chemokine
Gene Aliases: A-152E5.3; ABCD-2; CCL17; SCYA17; Scya17l; TARC
UniProt ID: (Human) Q92583
Entrez Gene ID: (Human) 6361, (Mouse) 20295
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.