Invitrogen
Manufacturer Code:PA5116367
Catalog # PIPA5116367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | recombinant protein of human CCL17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-152E5.3 ABCD-2 CC chemokine TARC C-C motif chemokine C-C motif chemokine 17 C-C motif chemokine ligand 17 CCL17 chemokine (C-C motif) ligand 17 Scya17 Scya17l small inducible cytokine subfamily A (Cys-Cys) member 17 small inducible cytokine subfamily A17 small-inducible cytokine A17 T cell-directed CC chemokine TARC Thymus and activation-regulated chemokine; C-C motif chemokine 17; CC chemokine TARC; chemokine (C-C motif) ligand 17; small inducible cytokine subfamily A (Cys-Cys), member 17; Small-inducible cytokine A17; T cell-directed CC chemokine; Thymus and activation-regulated chemokine
Gene Aliases: A-152E5.3; ABCD-2; CCL17; SCYA17; TARC
UniProt ID: (Human) Q92583
Entrez Gene ID: (Human) 6361
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.