R&D Systems
Manufacturer Code:MAB2432SP
Catalog # MAB2432SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 998524 |
Immunogen | Synthetic peptide containing human TAP2 Accession # Q03519 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: ABC transporter, MHC 2; ABC18 ABCB3 ABCB3ABC18 antigen peptide transporter 2 APT2 ATP-binding cassette sub-family B (MDR/TAP) member 3 D6S217E MHC 2 Peptide transporter involved in antigen processing 2 Peptide transporter PSF2 Peptide transporter TAP2 PSF2 PSF2PSF-2 Really interesting new gene 11 protein RING11 RING11ATP-binding cassette sub-family B member 3 transporter 2 ABC (ATP binding cassette) transporter 2 ATP-binding cassette sub-family B (MDR/TAP) Y1; Antigen peptide transporter 2; APT2; ATP-binding cassette sub-family B member 3; ATP-binding cassette, sub-family B (MDR/TAP), member 3; Peptide supply factor 2; Peptide transporter involved in antigen processing 2; Peptide transporter PSF2; Peptide transporter TAP2; PSF-2; Really interesting new gene 11 protein; RING11; transporter 2, ABC (ATP binding cassette); transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases: ABC18; ABCB3; APT2; D6S217E; PSF-2; PSF2; RING11; TAP2; Y1
UniProt ID: (Human) Q03519
Entrez Gene ID: (Human) 6891
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.