Invitrogen
Manufacturer Code:PA5102438
Catalog # PIPA5102438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAP2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC transporter MHC 2 ABC18 Abcb3 AI462429 Antigen peptide transporter 2 APT2 ATP dependent transport protein family member ATP-binding cassette sub-family B member 3 ATP-binding cassette sub-family B (MDR/TAP) member 3 Cim D6S217E Ham2 Ham-2 histocompatibility antigen modifier 2 jas MTP2 Peptide supply factor 2 Peptide transporter involved in antigen processing 2 Peptide transporter PSF2 Peptide transporter TAP2 PSF2 PSF-2 Really interesting new gene 11 protein RING11 Tap2 Tap-2 Transporter 2 ABC (ATP binding cassette) transporter 2 ATP-binding cassette sub-family B transporter 2 ABC (ATP binding cassette) transporter 2 ATP binding cassette subfamily B member transporter 2 ATP-binding cassette sub-family B (MDR/TAP) Y1; ABC transporter, MHC 2; Antigen peptide transporter 2; APT2; ATP dependent transport protein family member; ATP-binding cassette sub-family B member 3; ATP-binding cassette, sub-family B (MDR/TAP), member 3; Histocompatibility antigen modifier 2; Peptide supply factor 2; Peptide transporter involved in antigen processing 2; Peptide transporter PSF2; Peptide transporter TAP2; PSF-2; Really interesting new gene 11 protein; RING11; Transporter 2 ABC (ATP binding cassette); transporter 2 ATP-binding cassette sub-family B; transporter 2, ABC (ATP binding cassette); transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases: ABC18; ABCB3; AI462429; APT2; Cim; D6S217E; Ham-2; Ham2; jas; MTP2; PSF-2; PSF2; RING11; Tap-2; TAP2; Y1
UniProt ID: (Human) Q03519, (Mouse) P36371, (Rat) P36372
Entrez Gene ID: (Human) 6891, (Mouse) 21355, (Rat) 24812
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.