Invitrogen
Manufacturer Code:PA580094
Catalog # PIPA580094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human TAP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC transporter, MHC 1; Antigen peptide transporter 1; Antigen peptide transporter 1 APT1 ATP-binding cassette sub-family B member 2 Peptide supply factor 1 Peptide transporter PSF1 PSF-1 Peptide transporter TAP1 Peptide transporter involved in antigen processing 1 Really interesting new gene 4 protein TAP1 ABCB2 PSF1 RING4 Y3; APT1; ATP-binding cassette sub-family B member 2; ATP-binding cassette, sub-family B (MDR/TAP), member 2; Peptide supply factor 1; Peptide transporter involved in antigen processing 1; Peptide transporter PSF1; Peptide transporter TAP1; PSF-1; Really interesting new gene 4 protein; RING4; transporter 1 ATP-binding cassette sub-family B; transporter 1, ATP-binding cassette, sub-family B (MDR/TAP); transporter associated with antigen processing; transporter, ATP-binding cassette, major histocompatibility complex, 1
Gene Aliases: ABC17; ABCB2; APT1; D6S114E; PSF-1; PSF1; RING4; TAP1; TAP1*0102N; TAP1N; Y3
UniProt ID: (Human) Q03518
Entrez Gene ID: (Human) 6890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.