Invitrogen
Manufacturer Code:MA527220
Catalog # PIMA527220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E4 |
Immunogen | Full length human recombit protein of TALDO1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dihydroxyacetone transferase; glycerone transferase; TAL TAL-H TALDOR TALH dihydroxyacetone transferase glycerone transferase transaldolase; testicular secretory protein Li 56; Transaldolase
Gene Aliases: TAL; TAL-H; TALDO; TALDO1; TALDOR; TALH
UniProt ID: (Human) P37837
Entrez Gene ID: (Human) 6888
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.