Novus Biologicals
Manufacturer Code:NBP234783APC
Catalog # NBP234783J
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SPM536 |
Immunogen | Purified TAG-72 protein |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CA 72-4 Protein TAG-72 tag 72 tag-72 Protein TAG-72; mRNA (guanine-N(7)-)-methyltransferase; mRNA cap guanine-N7 methyltransferase
Gene Aliases: C25A1.3; CELE_C25A1.3; tag-72
UniProt ID: (C. elegans) Q9XVS1
Entrez Gene ID: (C. elegans) 182875
Molecular Function: RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.