Invitrogen
Manufacturer Code:PA5100758
Catalog # PIPA5100758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAF7L corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4933438I11Rik 50kDa Cancer/testis antigen 40 CT40 RNA polymerase II TBP-associated factor subunit Q Taf2q TAF7L TAF7-like RNA polymerase II TATA box binding protein (TBP)-associated factor TAF7-like RNA polymerase II TATA box binding protein (TBP)-associated factor 50kDa TATA box binding protein (TBP)-associated factor RNA polymerase II Q TATA box binding protein associated factor 7 like TATA box binding protein-associated factor RNA polymerase II subunit Q TATA box-binding protein-associated factor 50 kDa TATA-box binding protein associated factor 7 like TBP-associated factor RNA polymerase II Q transcription initiation factor TFIID 50 kDa subunit Transcription initiation factor TFIID subunit 7-like; Cancer/testis antigen 40; CT40; RNA polymerase II TBP-associated factor subunit Q; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, Q; TATA box binding protein associated factor 7 like; TATA box binding protein-associated factor RNA polymerase II subunit Q; TATA box-binding protein-associated factor 50 kDa; TBP-associated factor, RNA polymerase II, Q; Transcription initiation factor TFIID 50 kDa subunit; Transcription initiation factor TFIID subunit 7-like
Gene Aliases: 4933438I11Rik; 50kDa; CT40; TAF2Q; TAF7L
UniProt ID: (Human) Q5H9L4, (Mouse) Q9D3R9
Entrez Gene ID: (Human) 54457, (Mouse) 74469
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.