Novus Biologicals
Manufacturer Code:NBP184350
Catalog # NBP184350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ESQDEVPDEVENQFILRLPLEHACTVRNLARSQSVKMKDKLKIDLLPDGRHAVVEVEDVPLAAKLVDLPCVIESLRTLDKKTFYKTADISQMLVCTADGDIHLSPEEPAASTDPNIVRKKERGREEKCVWKHGITPPLKNVRK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50kDa CT40FLJ23157 RNA polymerase II TBP-associated factor subunit Q TAF2QRNA polymerase II Q TAF7-like RNA polymerase II TATA box binding protein (TBP)-associated factor TATA box binding protein (TBP)-associated factor RNA polymerase II Q TATA box-binding protein-associated factor 50 kDa transcription initiation factor TFIID subunit 7-like; Cancer/testis antigen 40; CT40; RNA polymerase II TBP-associated factor subunit Q; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa; TATA box binding protein associated factor 7 like; TATA box-binding protein-associated factor 50 kDa; TBP-associated factor, RNA polymerase II, Q; Transcription initiation factor TFIID 50 kDa subunit; Transcription initiation factor TFIID subunit 7-like
Gene Aliases: CT40; TAF2Q; TAF7L
UniProt ID: (Human) Q5H9L4
Entrez Gene ID: (Human) 54457
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.