Invitrogen
Manufacturer Code:PA528205
Catalog # PIPA528205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 288 of Human TAF7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RNA polymerase II TBP-associated factor subunit F; TAF(II)55; TAF2F TAFII55 RNA polymerase II TBP-associated factor subunit F TAF(II)55 TAFII-55 TATA box binding protein (TBP)-associated factor RNA polymerase II F 55kD TBP-associated factor F transcription factor IID subunit TAFII55 transcription initiation factor TFIID 55 kDa; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa; TAFII-55; TATA box binding protein (TBP)-associated factor, RNA polymerase II, F, 55kD; TATA box binding protein associated factor 7; TBP-associated factor F; transcription factor IID subunit TAFII55; Transcription initiation factor TFIID 55 kDa subunit; Transcription initiation factor TFIID subunit 7; transcription initiation factor TFIID, 55 kDa subunit
Gene Aliases: TAF2F; TAF7; TAFII55
UniProt ID: (Human) Q15545
Entrez Gene ID: (Human) 6879
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.