Invitrogen
Manufacturer Code:PA568520
Catalog # PIPA568520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat TAF6L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p300/CBP-associated factor (PCAF)-associated factor 65; PAF65-alpha; PCAF-associated factor 65 alpha; PCAF-associated factor 65-alpha; TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 6L; TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 6L PCAF-associated factor 65-alpha PAF65-alpha TAF6L PAF65A; TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa; TAF6L; TATA box binding protein associated factor 6 like
Gene Aliases: 2810417N14Rik; BB223262; C530024J06Rik; fa05b10; PAF65A; TAF6L; wu:fa05b10
UniProt ID: (Human) Q9Y6J9, (Mouse) Q8R2K4
Entrez Gene ID: (Human) 10629, (Bovine) 531109, (Rabbit) 100339101, (Dog) 476051, (Horse) 100063916, (Mouse) 225895, (Guinea pig) 100733115, (Rat) 309194, (Zebrafish) 557363
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.