Invitrogen
Manufacturer Code:MA3077
Catalog # PIMA3077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 25TA-2G7 |
Immunogen | Synthetic peptide coupled to ovalalbumin from aa 39 to aa 56 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: p80; RNA polymerase II TBP-associated factor subunit E; RNA polymerase II TBP-associated factor subunit E TATA box binding protein (TBP)-associated factor transcription initiation factor TFIID 70 kDa subunit transcription initiation factor TFIID subunit 6 TAF(II)70 TATA box binding protein (TBP)-associated factor 80 kDa TAFII-70 TAFII-80 TAFII80 transcription initiation factor TFIID 80 kDa subunit TAF2E TAFII70 TAF(II)80; TAF(II)70; TAF(II)80; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80 kDa; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAFII-70; TAFII-80; TAFII80; TATA box binding protein (Tbp)-associated factor, RNA polymerase II, E; TATA box binding protein (TBP)-associated factor, RNA polymerase II, E, 70/85kD; TATA box binding protein associated factor 6; Transcription initiation factor TFIID 70 kDa subunit; Transcription initiation factor TFIID 80 kDa subunit; Transcription initiation factor TFIID subunit 6
Gene Aliases: 80kDa; AW549759; MGC:8964; p80; TAF(II)70; TAF(II)80; TAF2E; TAF6; TAFII-70; TAFII-80; TAFII70; TAFII80; TAFII85
UniProt ID: (Human) P49848, (Mouse) Q62311
Entrez Gene ID: (Human) 6878, (Mouse) 21343
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.