Novus Biologicals
Manufacturer Code:NBP295182100UL
Catalog # NB024526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 350-450 of human TAF6 (XP_006716164.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781E21155 MGC:8964 RNA polymerase II E 70/85kD TAF(II)70 TAF(II)80 TAF2ERNA polymerase II TBP-associated factor subunit E TAF6 RNA polymerase II TATA box binding protein (TBP)-associated factor 80kDa TAFII70TAFII-80 TAFII80TAFII-70 TAFII85 Transcription initiation factor TFIID 70 kDa subunit Transcription initiation factor TFIID 80 kDa subunit transcription initiation factor TFIID subunit 6; RNA polymerase II TBP-associated factor subunit E; TAF(II)70; TAF(II)80; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, E, 70/85kD; TATA box binding protein associated factor 6; Transcription initiation factor TFIID 70 kDa subunit; Transcription initiation factor TFIID 80 kDa subunit; Transcription initiation factor TFIID subunit 6
Gene Aliases: MGC:8964; TAF(II)70; TAF(II)80; TAF2E; TAF6; TAFII-70; TAFII-80; TAFII70; TAFII80; TAFII85
UniProt ID: (Human) P49848
Entrez Gene ID: (Human) 6878
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.