Abnova
Manufacturer Code:H00027097M01A
Catalog # 89-106-741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C5 |
Immunogen | TAF5L (NP_0552241 a.a. &bsim 117 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant TAF5L.
For Research Use Only
Protein Aliases: PAF65-beta; PAF65B; PCAF associated factor 65 beta; PCAF-associated factor 65 beta; TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L; TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa; TAF5L; TATA box binding protein associated factor 5 like
Gene Aliases: PAF65B; TAF5L
UniProt ID: (Human) O75529
Entrez Gene ID: (Human) 27097
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.