Invitrogen
Manufacturer Code:PA536309
Catalog # PIPA536309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 301-350 of human TAF5L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAF65-beta; PCAF associated factor 65 beta; PCAF-associated factor 65 beta; TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L; TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L PCAF-associated factor 65 beta PAF65-beta TAF5L PAF65B; TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa; TAF5L; TATA box binding protein associated factor 5 like
Gene Aliases: 1110005N04Rik; AI849020; PAF65B; TAF5L
UniProt ID: (Human) O75529, (Mouse) Q91WQ5
Entrez Gene ID: (Human) 27097, (Mouse) 102162, (Rat) 307927
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.