Novus Biologicals
Manufacturer Code:NBP293025100UL
Catalog # NB027945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 656-800 of human TAF5 (NP_008882.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TAF(II)100; TAF(II)100 TAF2DTATA box binding protein (TBP)-associated factor 2D TAF5 RNA polymerase II TATA box binding protein (TBP)-associated factor TAFII100TAFII-100 TATA box binding protein (TBP)-associated factor RNA polymerase II D 100kD transcription initiation factor TFIID 100 kD subunit Transcription initiation factor TFIID 100 kDa subunit transcription initiation factor TFIID subunit 5; TAF5 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 100kDa; TATA box binding protein (TBP)-associated factor 2D; TATA box binding protein (TBP)-associated factor, RNA polymerase II, D, 100kD; TATA box binding protein associated factor 5; transcription initiation factor TFIID 100 kD subunit; Transcription initiation factor TFIID 100 kDa subunit; Transcription initiation factor TFIID subunit 5
Gene Aliases: TAF(II)100; TAF2D; TAF5; TAFII-100; TAFII100
UniProt ID: (Human) Q15542
Entrez Gene ID: (Human) 6877
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.